siRNA Details
| Gene Symbol: Fam89b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015013.1 | NP_001015013.1 | 2 | 751 | ||
| GenBank mRNA | BC093377.1 | - | 2 | 751 | 7388027 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam89b| Gene Symbol: Fam89b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015013.1 | NP_001015013.1 | 2 | 751 | ||
| GenBank mRNA | BC093377.1 | - | 2 | 751 | 7388027 | |