siRNA Details
| Gene Symbol: LOC314140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009694.1 | NP_001009694.1 | 2 | 1573 | ||
| XM_008764657.1 | XP_008762879.1 | 1 | 1349 | |||
| GenBank mRNA | BC088149.1 | - | 2 | 1573 | 7380332 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC314140| Gene Symbol: LOC314140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009694.1 | NP_001009694.1 | 2 | 1573 | ||
| XM_008764657.1 | XP_008762879.1 | 1 | 1349 | |||
| GenBank mRNA | BC088149.1 | - | 2 | 1573 | 7380332 | |