siRNA Details
| Gene Symbol: Zfp422 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012745.1 | NP_001012763.1 | 2 | 1657 | ||
| XM_006237187.2 | XP_006237249.1 | 2 | 1649 | |||
| XM_006237189.2 | XP_006237251.1 | 2 | 1477 | |||
| XM_006237190.2 | XP_006237252.1 | 2 | 1508 | |||
| XM_008763249.1 | XP_008761471.1 | 2 | 1585 | |||
| GenBank mRNA | AY724498.1 | - | 2 | 1479 | ||
| BC090354.1 | - | 2 | 1657 | 7319307 | ||

Zfp422
Genomic Map