siRNA Details
| Gene Symbol: Rnase17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012232.1 | NP_001012232.1 | 1 | 181 | ||
| XM_008770561.1 | XP_008768783.1 | 2 | 259 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnase17| Gene Symbol: Rnase17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012232.1 | NP_001012232.1 | 1 | 181 | ||
| XM_008770561.1 | XP_008768783.1 | 2 | 259 | |||