siRNA Details
Gene Symbol: LOC502176 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025057.2 | NP_001020228.2 | 2, 3 | 213 | ||
GenBank mRNA | AY325157.1 | - | 2, 3 | 213 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC502176 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025057.2 | NP_001020228.2 | 2, 3 | 213 | ||
GenBank mRNA | AY325157.1 | - | 2, 3 | 213 |