siRNA Details
| Gene Symbol: Cacna1h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153814.2 | NP_722521.1 | 34 | 6642 | ||
| GenBank mRNA | AF290213.2 | - | 34 | 6645 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cacna1h| Gene Symbol: Cacna1h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153814.2 | NP_722521.1 | 34 | 6642 | ||
| GenBank mRNA | AF290213.2 | - | 34 | 6645 | ||