siRNA Details
| Gene Symbol: Sfpq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025271.1 | NP_001020442.1 | 9 | 1987 | ||
| GenBank mRNA | AF036335.1 | - | 5 | 517 | ||
| BC098731.1 | - | 9 | 1987 | 7384804 | ||
| FQ227268.1 | - | 5 | 452 | |||
| FQ229853.1 | - | 5 | 529 | |||

Sfpq
Genomic Map