siRNA Details
| Gene Symbol: Sfpq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025271.1 | NP_001020442.1 | 5 | 1647 | ||
| GenBank mRNA | AF036335.1 | - | 1 | 176 | ||
| BC098731.1 | - | 5 | 1647 | 7384804 | ||
| FQ227268.1 | - | 1 | 112 | |||
| FQ229853.1 | - | 1 | 189 | |||

Sfpq
Genomic Map