siRNA Details
| Gene Symbol: Smndc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025400.3 | NP_001020571.1 | 6 | 848 | ||
| NM_001035234.2 | NP_001030311.1 | 6 | 1010 | |||
| XM_006231608.2 | XP_006231670.1 | 6 | 1287 | |||
| GenBank mRNA | BC097986.1 | - | 6 | 848 | 7444662 | |
| BC098697.1 | - | 6 | 989 | 7386640 | ||
| FQ220088.1 | - | 5 | 791 | |||

Smndc1
Genomic Map