siRNA Details
| Gene Symbol: Umps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025402.1 | NP_001020573.1 | 3 | 606 | ||
| GenBank mRNA | BC098033.1 | - | 3 | 606 | 7456468 | |
| FQ233727.1 | - | 2 | 344 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Umps| Gene Symbol: Umps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025402.1 | NP_001020573.1 | 3 | 606 | ||
| GenBank mRNA | BC098033.1 | - | 3 | 606 | 7456468 | |
| FQ233727.1 | - | 2 | 344 | |||