siRNA Details
| Gene Symbol: Mospd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025629.1 | NP_001020800.1 | 2 | 349 | ||
| XM_006249115.2 | XP_006249177.1 | 3 | 409 | |||
| GenBank mRNA | BC099230.1 | - | 2 | 349 | 7442803 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mospd3| Gene Symbol: Mospd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025629.1 | NP_001020800.1 | 2 | 349 | ||
| XM_006249115.2 | XP_006249177.1 | 3 | 409 | |||
| GenBank mRNA | BC099230.1 | - | 2 | 349 | 7442803 | |