siRNA Details
| Gene Symbol: FMN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001305424.1 | NP_001292353.1 | 14 | 4946 | ||
| NM_020066.4 | NP_064450.3 | 13 | 4934 | |||
| XM_017001839.1 | XP_016857328.1 | 14 | 3786 | |||
| XM_017001840.1 | XP_016857329.1 | 13 | 3186 | |||
| XM_017001841.1 | XP_016857330.1 | 13 | 3644 | |||
| XM_017001842.1 | XP_016857331.1 | 12 | 2865 | |||
| XM_017001843.1 | XP_016857332.1 | 11 | 2853 | |||
| GenBank mRNA | AB209153.1 | - | 14 | 3539 | ||
| AF218942.1 | - | 5 | 434 | |||
| AF225426.1 | - | 5 | 456 | |||
| AK297716.1 | - | 8 | 704 | |||
| AK297755.1 | - | 8 | 950 | |||
| AK298141.1 | - | 6 | 769 | |||
| BC014364.2 | - | 3 | 273 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_019445, AK013585, AF218940, BC094606, AK140268.

FMN2
Genomic Map