siRNA Details
Gene Symbol: Ewsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025632.2 | NP_001020803.2 | 17 | 2143 | ||
XM_006251201.2 | XP_006251263.2 | 17 | 2128 | |||
XM_006251202.2 | XP_006251264.2 | 17 | 2125 | |||
XM_006251203.2 | XP_006251265.2 | 17 | 2154 | |||
GenBank mRNA | BC098822.1 | - | 8 | 1177 | ||
BC162004.1 | - | 17 | 2143 | 9089248 | ||
FQ221459.1 | - | 8 | 1290 |
Gene Symbol: LOC100912481 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749683.2 | XP_003749731.1 | 17 | 2148 | ||
XM_003749684.2 | XP_003749732.1 | 18 | 2163 | |||
XM_006236016.1 | XP_006236078.1 | 18 | 2166 | |||
XM_006236017.1 | XP_006236079.1 | 17 | 2145 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001283062, NM_001283061, NM_007968, AK147103, BC068226, and others.
Panther Classification:
Gene Ontology Categories:
