siRNA Details
| Gene Symbol: Vnn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025623.1 | NP_001020794.1 | 7 | 2211 | ||
| GenBank mRNA | BC098934.1 | - | 7 | 2211 | 7441890 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vnn1| Gene Symbol: Vnn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025623.1 | NP_001020794.1 | 7 | 2211 | ||
| GenBank mRNA | BC098934.1 | - | 7 | 2211 | 7441890 | |