siRNA Details
| Gene Symbol: Psmb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025637.1 | NP_001020808.1 | 8 | 974 | ||
| GenBank mRNA | BC098835.1 | - | 8 | 974 | 7378841 | |
| FQ217478.1 | - | 4 | 517 | |||
| FQ226463.1 | - | 8 | 967 | |||
| FQ228226.1 | - | 8 | 954 | |||
| FQ229295.1 | - | 8 | 972 | |||
| FQ233484.1 | - | 8 | 984 | |||
| FQ234080.1 | - | 8 | 976 | |||
| FQ235075.1 | - | 8 | 957 | |||

Psmb10
Genomic Map