siRNA Details
| Gene Symbol: Psmb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025637.1 | NP_001020808.1 | 3 | 328 | ||
| GenBank mRNA | BC098835.1 | - | 3 | 328 | 7378841 | |
| FQ226463.1 | - | 3 | 321 | |||
| FQ228226.1 | - | 3 | 309 | |||
| FQ229295.1 | - | 3 | 326 | |||
| FQ229779.1 | - | 381 | ||||
| FQ233484.1 | - | 3 | 338 | |||
| FQ234080.1 | - | 3 | 330 | |||
| FQ235075.1 | - | 3 | 311 | |||

Psmb10
Genomic Map