siRNA Details
| Gene Symbol: Vta1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025640.1 | NP_001020811.1 | 4, 5 | 423 | ||
| XM_006227646.2 | XP_006227708.1 | 5, 6 | 573 | |||
| GenBank mRNA | BC098787.1 | - | 4, 5 | 423 | 7375248 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vta1| Gene Symbol: Vta1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025640.1 | NP_001020811.1 | 4, 5 | 423 | ||
| XM_006227646.2 | XP_006227708.1 | 5, 6 | 573 | |||
| GenBank mRNA | BC098787.1 | - | 4, 5 | 423 | 7375248 | |