siRNA Details
Gene Symbol: Ceacam11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025404.1 | NP_001020575.1 | 3 | 637 | ||
GenBank mRNA | BC097990.1 | - | 3 | 637 | 7445748 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ceacam11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025404.1 | NP_001020575.1 | 3 | 637 | ||
GenBank mRNA | BC097990.1 | - | 3 | 637 | 7445748 |