siRNA Details
| Gene Symbol: Poc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025647.1 | NP_001020818.1 | 13 | 2040 | ||
| XM_006231821.1 | XP_006231883.1 | 11 | 1736 | |||
| XM_008760664.1 | XP_008758886.1 | 10 | 1781 | |||
| XM_008760665.1 | XP_008758887.1 | 11 | 1659 | |||
| GenBank mRNA | BC097489.1 | - | 13 | 2040 | 7460568 | |

Poc5
Genomic Map