siRNA Details
Gene Symbol: Snapin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025648.1 | NP_001020819.1 | 2 | 654 | ||
NM_001170576.1 | NP_001164047.1 | 4 | 571 | |||
GenBank mRNA | BC098864.1 | - | 2 | 654 | 7380523 | |
DY315040.1 | - | 4 | 576 | |||
FQ218987.1 | - | 2 | 261 |
