siRNA Details
| Gene Symbol: Dusp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025650.1 | NP_001020821.1 | 7, 8 | 670 | ||
| GenBank mRNA | BC098712.1 | - | 7, 8 | 670 | 7321787 | |
| Gene Symbol: LOC103690006 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763101.1 | XP_008761323.1 | 7, 8 | 749 | ||

Dusp11
Genomic Map