siRNA Details
| Gene Symbol: Sytl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025651.1 | NP_001020822.1 | 14 | 1714 | ||
| GenBank mRNA | BC098679.1 | - | 14 | 1714 | 7384784 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sytl1| Gene Symbol: Sytl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025651.1 | NP_001020822.1 | 14 | 1714 | ||
| GenBank mRNA | BC098679.1 | - | 14 | 1714 | 7384784 | |