siRNA Details
| Gene Symbol: Cmpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025655.1 | NP_001020826.1 | 6 | 1001 | ||
| GenBank mRNA | BC098727.1 | - | 6 | 1001 | 7379366 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cmpk1| Gene Symbol: Cmpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025655.1 | NP_001020826.1 | 6 | 1001 | ||
| GenBank mRNA | BC098727.1 | - | 6 | 1001 | 7379366 | |