siRNA Details
| Gene Symbol: Ssu72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025657.1 | NP_001020828.1 | 1, 2 | 351 | ||
| GenBank mRNA | BC099142.1 | - | 1, 2 | 351 | 7300601 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ssu72| Gene Symbol: Ssu72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025657.1 | NP_001020828.1 | 1, 2 | 351 | ||
| GenBank mRNA | BC099142.1 | - | 1, 2 | 351 | 7300601 | |