siRNA Details
Gene Symbol: Ei24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025660.1 | NP_001020831.1 | 12 | 1907 | ||
XM_006242800.2 | XP_006242862.1 | 11 | 1998 | |||
XM_006242801.2 | XP_006242863.1 | 11 | 1955 | |||
GenBank mRNA | BC098721.1 | - | 12 | 1907 | 7370909 |
