siRNA Details
| Gene Symbol: Dnaja4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025411.1 | NP_001020582.1 | 6 | 1267 | ||
| GenBank mRNA | BC082010.1 | - | 6 | 1265 | 7131770 | |
| BC097438.1 | - | 6 | 1267 | 7455678 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnaja4| Gene Symbol: Dnaja4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025411.1 | NP_001020582.1 | 6 | 1267 | ||
| GenBank mRNA | BC082010.1 | - | 6 | 1265 | 7131770 | |
| BC097438.1 | - | 6 | 1267 | 7455678 | ||