siRNA Details
Gene Symbol: Fam58b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025412.1 | NP_001020583.1 | 1 | 930 | ||
GenBank mRNA | BC086445.1 | - | 1 | 917 | ||
BC097941.1 | - | 1 | 930 | 7376750 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
