siRNA Details
| Gene Symbol: Tmem184a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025413.1 | NP_001020584.1 | 7 | 1000 | ||
| XM_006248938.2 | XP_006249000.1 | 8 | 1196 | |||
| XM_006248939.2 | XP_006249001.1 | 8 | 1188 | |||
| XM_006248940.2 | XP_006249002.1 | 8 | 1103 | |||
| XM_008768972.1 | XP_008767194.1 | 8 | 1012 | |||
| GenBank mRNA | BC083910.1 | - | 7 | 998 | ||
| BC098056.1 | - | 7 | 1000 | 7457725 | ||

Tmem184a
Genomic Map