siRNA Details
| Gene Symbol: Tmem184a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025413.1 | NP_001020584.1 | 8 | 1128 | ||
| XM_006248938.2 | XP_006249000.1 | 9 | 1324 | |||
| XM_006248939.2 | XP_006249001.1 | 9 | 1316 | |||
| XM_006248940.2 | XP_006249002.1 | 9 | 1231 | |||
| XM_008768972.1 | XP_008767194.1 | 9 | 1140 | |||
| GenBank mRNA | BC083910.1 | - | 8 | 1126 | ||
| BC098056.1 | - | 8 | 1128 | 7457725 | ||

Tmem184a
Genomic Map