siRNA Details
| Gene Symbol: Mgat4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025666.1 | NP_001020837.1 | 2 | 262 | ||
| XM_008772379.1 | XP_008770601.1 | 2 | 347 | |||
| XM_008772380.1 | XP_008770602.1 | 2 | 199 | |||
| GenBank mRNA | BC097408.1 | - | 2 | 262 | 7454086 | |

Mgat4d
Genomic Map