siRNA Details
| Gene Symbol: Rnf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025667.1 | NP_001020838.1 | 4 | 569 | ||
| XM_006249990.2 | XP_006250052.1 | 4 | 507 | |||
| XM_006249991.2 | XP_006250053.1 | 4 | 589 | |||
| GenBank mRNA | AB568264.1 | - | 3 | 373 | ||
| BC098941.1 | - | 4 | 569 | 7456372 | ||

Rnf2
Genomic Map