siRNA Details
| Gene Symbol: Rassf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025671.1 | NP_001020842.1 | 3 | 287 | ||
| XM_006250745.2 | XP_006250807.1 | 4 | 399 | |||
| XM_006250747.2 | XP_006250809.1 | 4 | 387 | |||
| XM_006250748.1 | XP_006250810.1 | 4 | 422 | |||
| XM_008770036.1 | XP_008768258.1 | 4 | 482 | |||
| XM_008770037.1 | XP_008768259.1 | 4 | 432 | |||
| XM_008770038.1 | XP_008768260.1 | 4 | 3674 | |||
| GenBank mRNA | BC097375.1 | - | 3 | 287 | 7444629 | |

Rassf6
Genomic Map