siRNA Details
| Gene Symbol: Rbm22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025676.1 | NP_001020847.1 | 4 | 210 | ||
| GenBank mRNA | BC097991.1 | - | 4 | 210 | 7446014 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbm22| Gene Symbol: Rbm22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025676.1 | NP_001020847.1 | 4 | 210 | ||
| GenBank mRNA | BC097991.1 | - | 4 | 210 | 7446014 | |