siRNA Details
| Gene Symbol: Hars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025414.1 | NP_001020585.1 | 3 | 282 | ||
| GenBank mRNA | BC097969.1 | - | 3 | 282 | 7442005 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hars| Gene Symbol: Hars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025414.1 | NP_001020585.1 | 3 | 282 | ||
| GenBank mRNA | BC097969.1 | - | 3 | 282 | 7442005 | |