siRNA Details
Gene Symbol: Cdr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025682.1 | NP_001020853.1 | 3 | 317 | ||
GenBank mRNA | BC098028.1 | - | 3 | 317 | 7456103 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cdr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025682.1 | NP_001020853.1 | 3 | 317 | ||
GenBank mRNA | BC098028.1 | - | 3 | 317 | 7456103 |