siRNA Details
| Gene Symbol: RGD1311946 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025683.1 | NP_001020854.1 | 4 | 2043 | ||
| GenBank mRNA | BC098053.1 | - | 4 | 2043 | 7457583 | |
| FQ229270.1 | - | 1 | 1014 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1311946| Gene Symbol: RGD1311946 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025683.1 | NP_001020854.1 | 4 | 2043 | ||
| GenBank mRNA | BC098053.1 | - | 4 | 2043 | 7457583 | |
| FQ229270.1 | - | 1 | 1014 | |||