siRNA Details
| Gene Symbol: Cd5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025685.1 | NP_001020856.1 | 6 | 1755 | ||
| GenBank mRNA | BC098724.1 | - | 6 | 1755 | 7378264 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd5l| Gene Symbol: Cd5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025685.1 | NP_001020856.1 | 6 | 1755 | ||
| GenBank mRNA | BC098724.1 | - | 6 | 1755 | 7378264 | |