siRNA Details
| Gene Symbol: Psmd14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025689.1 | NP_001020860.1 | 8 | 809 | ||
| GenBank mRNA | BC097427.1 | - | 8 | 809 | ||
| BC158645.1 | - | 8 | 757 | 8363877 | ||
| FQ228714.1 | - | 8 | 785 | |||
| FQ228827.1 | - | 8 | 785 | |||

Psmd14
Genomic Map