siRNA Details
| Gene Symbol: Sdcbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025692.1 | NP_001020863.1 | 7 | 692 | ||
| XM_008762325.1 | XP_008760547.1 | 5 | 2668 | |||
| XM_008762326.1 | XP_008760548.1 | 8 | 1673 | |||
| GenBank mRNA | BC099095.1 | - | 7 | 692 | 7459023 | |

Sdcbp2
Genomic Map