siRNA Details
Gene Symbol: Cdca7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025693.1 | NP_001020864.1 | 8 | 1176 | ||
GenBank mRNA | BC098690.1 | - | 8 | 1176 | 7386103 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cdca7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025693.1 | NP_001020864.1 | 8 | 1176 | ||
GenBank mRNA | BC098690.1 | - | 8 | 1176 | 7386103 |