siRNA Details
| Gene Symbol: Sf3a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025698.1 | NP_001020869.1 | 1 | 120 | ||
| GenBank mRNA | AY387079.1 | - | 1 | 57 | ||
| BC099183.1 | - | 1 | 120 | 7383264 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sf3a3| Gene Symbol: Sf3a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025698.1 | NP_001020869.1 | 1 | 120 | ||
| GenBank mRNA | AY387079.1 | - | 1 | 57 | ||
| BC099183.1 | - | 1 | 120 | 7383264 | ||