siRNA Details
| Gene Symbol: Phf20l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765536.1 | XP_008763758.1 | 9 | 1763 | ||
| XM_008765537.1 | XP_008763759.1 | 7 | 1608 | |||
| GenBank mRNA | BC096896.1 | - | 9 | 1763 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phf20l1| Gene Symbol: Phf20l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765536.1 | XP_008763758.1 | 9 | 1763 | ||
| XM_008765537.1 | XP_008763759.1 | 7 | 1608 | |||
| GenBank mRNA | BC096896.1 | - | 9 | 1763 | ||