siRNA Details
| Gene Symbol: Azi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025705.1 | NP_001020876.1 | 8 | 1921 | ||
| XM_006244032.2 | XP_006244094.1 | 9 | 2792 | |||
| GenBank mRNA | BC098680.1 | - | 8 | 1921 | 7384893 | |
| FQ221679.1 | - | 9 | 2041 | |||

Azi2
Genomic Map