siRNA Details
| Gene Symbol: Fem1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025706.1 | NP_001020877.1 | 1 | 951 | ||
| GenBank mRNA | BC097490.1 | - | 1 | 951 | 7460827 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fem1a| Gene Symbol: Fem1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025706.1 | NP_001020877.1 | 1 | 951 | ||
| GenBank mRNA | BC097490.1 | - | 1 | 951 | 7460827 | |