siRNA Details
| Gene Symbol: Ogfrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025708.1 | NP_001020879.1 | 5 | 630 | ||
| XM_008766933.1 | XP_008765155.1 | 5 | 678 | |||
| GenBank mRNA | BC099205.1 | - | 5 | 630 | 7384201 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ogfrl1| Gene Symbol: Ogfrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025708.1 | NP_001020879.1 | 5 | 630 | ||
| XM_008766933.1 | XP_008765155.1 | 5 | 678 | |||
| GenBank mRNA | BC099205.1 | - | 5 | 630 | 7384201 | |