siRNA Details
| Gene Symbol: Vps4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025716.1 | NP_001020887.1 | 10, 11 | 1414 | ||
| GenBank mRNA | BC099128.1 | - | 10, 11 | 1414 | 7459593 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vps4b| Gene Symbol: Vps4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025716.1 | NP_001020887.1 | 10, 11 | 1414 | ||
| GenBank mRNA | BC099128.1 | - | 10, 11 | 1414 | 7459593 | |