siRNA Details
| Gene Symbol: Arpc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025717.1 | NP_001020888.1 | 4 | 970 | ||
| GenBank mRNA | BC099238.1 | - | 4 | 970 | 7445509 | |
| FQ225700.1 | - | 2 | 594 | |||
| FQ230401.1 | - | 2 | 594 | |||
| FQ230694.1 | - | 1 | 190 | |||

Arpc5
Genomic Map