siRNA Details
| Gene Symbol: Tfdp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025718.1 | NP_001020889.1 | 9 | 1932 | ||
| XM_006253451.2 | XP_006253513.1 | 10 | 2197 | |||
| XM_008771397.1 | XP_008769619.1 | 11 | 2200 | |||
| XM_008771398.1 | XP_008769620.1 | 11 | 2279 | |||
| XM_008771399.1 | XP_008769621.1 | 11 | 2107 | |||
| XM_008771400.1 | XP_008769622.1 | 10 | 2233 | |||
| GenBank mRNA | BC098789.1 | - | 9 | 1932 | 7375514 | |

Tfdp1
Genomic Map