siRNA Details
| Gene Symbol: Cdk10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025722.2 | NP_001020893.2 | 9 | 1205 | ||
| NM_001109936.1 | NP_001103406.1 | 10 | 754 | |||
| NM_001109937.1 | NP_001103407.1 | 10 | 715 | |||
| GenBank mRNA | BC098804.1 | - | 9 | 1205 | 7376321 | |

Cdk10
Genomic Map