siRNA Details
| Gene Symbol: Mtrf1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025723.1 | NP_001020894.1 | 4, 5 | 729 | ||
| GenBank mRNA | BC097970.1 | - | 4, 5 | 729 | 7442024 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mtrf1l| Gene Symbol: Mtrf1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025723.1 | NP_001020894.1 | 4, 5 | 729 | ||
| GenBank mRNA | BC097970.1 | - | 4, 5 | 729 | 7442024 | |